Assay Details
Gene Symbol: mIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143364.3 | NP_651621.2 | 4 - 5 | 1343 | 78 | |
GenBank mRNA | BT150258.1 | - | 4 - 5 | 1320 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
