Assay Details
Gene Symbol: alph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260427.2 | NP_001247356.1 | 2 - 3 | 1234 | 86 | |
NM_143444.4 | NP_651701.1 | 2 - 3 | 1234 | 86 | ||
NM_170416.3 | NP_733295.1 | 2 - 3 | 1234 | 86 | ||
NM_170417.3 | NP_733296.1 | 2 - 3 | 999 | 86 | ||
NM_170418.3 | NP_733297.1 | 1 - 2 | 1075 | 86 | ||
NM_170419.3 | NP_733298.1 | 2 - 3 | 1234 | 86 | ||
GenBank mRNA | AY051685.1 | - | 2 - 3 | 1210 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
