Assay Details
Gene Symbol: alph | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260427.2 | NP_001247356.1 | 5 - 6 | 1539 | 92 | |
NM_143444.4 | NP_651701.1 | 4 - 5 | 1455 | 92 | ||
NM_170416.3 | NP_733295.1 | 4 - 5 | 1455 | 92 | ||
NM_170417.3 | NP_733296.1 | 4 - 5 | 1220 | 92 | ||
NM_170418.3 | NP_733297.1 | 3 - 4 | 1296 | 92 | ||
NM_170419.3 | NP_733298.1 | 4 - 5 | 1455 | 92 | ||
GenBank mRNA | AY051685.1 | - | 4 - 5 | 1431 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
