Assay Details
| Gene Symbol: Obp99b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276149.1 | NP_001263078.1 | 2 | 526 | 67 | |
| NM_143456.3 | NP_651713.1 | 1 | 514 | 67 | ||
| GenBank mRNA | AY119044.1 | - | 1 | 490 | 67 | |
More Information
Set Membership: |
|

Obp99b
Genomic Map