Assay Details
Gene Symbol: RpS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143525.4 | NP_651782.1 | 2 - 3 | 202 | 59 | |
NM_170476.2 | NP_733355.1 | 1 - 2 | 455 | 59 | ||
NM_170477.3 | NP_733356.2 | 1 - 2 | 207 | 59 | ||
NM_206589.2 | NP_996312.1 | 2 - 3 | 274 | 59 | ||
GenBank mRNA | AY071156.1 | - | 2 - 3 | 274 | 59 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
