Assay Details
Gene Symbol: RpS7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143525.4 | NP_651782.1 | 3 - 4 | 535 | 68 | |
NM_170476.2 | NP_733355.1 | 2 - 3 | 788 | 68 | ||
NM_170477.3 | NP_733356.2 | 2 - 3 | 540 | 68 | ||
NM_206589.2 | NP_996312.1 | 3 - 4 | 607 | 68 | ||
GenBank mRNA | AY071156.1 | - | 3 - 4 | 607 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
