Assay Details
| Gene Symbol: sip3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276223.2 | NP_001263152.1 | 1 - 2 | 459 | 87 | |
| NM_143637.5 | NP_651894.3 | 1 - 2 | 459 | 87 | ||
| GenBank mRNA | AY060677.1 | - | 1 - 2 | 440 | 87 | |
More Information
Set Membership: |
|

sip3
Genomic Map