Assay Details
Gene Symbol: GluRIID | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143725.3 | NP_651982.1 | 12 - 13 | 2545 | 70 | |
GenBank mRNA | AJ251888.1 | - | 12 - 13 | 2502 | 70 | |
AJ251889.1 | - | 12 - 13 | 2502 | 70 | ||
BT150368.1 | - | 12 - 13 | 2545 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
