Assay Details
| Gene Symbol: GluRIID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143725.3 | NP_651982.1 | 3 - 4 | 657 | 75 | |
| GenBank mRNA | AJ251888.1 | - | 3 - 4 | 614 | 75 | |
| AJ251889.1 | - | 3 - 4 | 614 | 75 | ||
| BT150368.1 | - | 3 - 4 | 657 | 75 | ||
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 71 and 85  Drosophila melanogaster | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 GluRIID
GluRIID Genomic Map
Genomic Map Back To Top
 Back To Top