Assay Details
| Gene Symbol: GluRIID | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143725.3 | NP_651982.1 | 4 - 5 | 1045 | 90 | |
| GenBank mRNA | AJ251888.1 | - | 4 - 5 | 1002 | 90 | |
| AJ251889.1 | - | 4 - 5 | 1002 | 90 | ||
| BT150368.1 | - | 4 - 5 | 1045 | 90 | ||
More Information
Set Membership: |
|

GluRIID
Genomic Map