Assay Details
| Gene Symbol: Lk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143729.3 | NP_651986.2 | 1 - 2 | 972 | 62 | |
| GenBank mRNA | BT050542.1 | - | 1 - 2 | 297 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lk6| Gene Symbol: Lk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143729.3 | NP_651986.2 | 1 - 2 | 972 | 62 | |
| GenBank mRNA | BT050542.1 | - | 1 - 2 | 297 | 62 | |
Set Membership: |
|