Assay Details
| Gene Symbol: Lk6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_143729.3 | NP_651986.2 | 4 - 5 | 1887 | 56 | |
| NM_169423.3 | NP_731624.1 | 4 - 5 | 1439 | 56 | ||
| GenBank mRNA | BT050542.1 | - | 4 - 5 | 1212 | 56 | |
More Information
Set Membership: |
|

Lk6
Genomic Map