Assay Details
Gene Symbol: Invadolysin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143815.4 | NP_652072.1 | 2 - 3 | 1005 | 82 | |
GenBank mRNA | AF234630.1 | - | 2 - 3 | 272 | 82 | |
BT001861.1 | - | 2 - 3 | 974 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
