Assay Details
Gene Symbol: Invadolysin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143815.4 | NP_652072.1 | 4 - 5 | 1116 | 87 | |
GenBank mRNA | AF234630.1 | - | 4 - 5 | 383 | 87 | |
BT001861.1 | - | 4 - 5 | 1085 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
