Assay Details
Gene Symbol: Invadolysin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_143815.4 | NP_652072.1 | 7 - 8 | 2036 | 83 | |
GenBank mRNA | AF234630.1 | - | 7 - 8 | 1303 | 83 | |
BT001861.1 | - | 7 - 8 | 2005 | 83 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
