Assay Details
| Gene Symbol: CG14645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144068.3 | NP_652325.2 | 1 | 279 | 61 | |
| GenBank mRNA | BT031033.1 | - | 1 | 279 | 61 | |
| Gene Symbol: RpL36A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BT029953.1 | - | 2078 | 61 | ||
| Gene Symbol: Skp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_164340.4 | NP_730815.2 | 5 | 1915 | 61 | |
More Information
Set Membership: |
|

CG14645
Genomic Map