Assay Details
| Gene Symbol: Snr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169055.3 | NP_730935.1 | 1 - 2 | 404 | 78 | |
| GenBank mRNA | AY070501.1 | - | 1 - 2 | 370 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Snr1| Gene Symbol: Snr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169055.3 | NP_730935.1 | 1 - 2 | 404 | 78 | |
| GenBank mRNA | AY070501.1 | - | 1 - 2 | 370 | 78 | |
Set Membership: |
|