Assay Details
| Gene Symbol: Itpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300251.1 | NP_001287180.1 | 12 - 13 | 5129 | 63 | |
| NM_169060.2 | NP_730941.1 | 13 - 14 | 5111 | 63 | ||
| NM_169061.2 | NP_730942.1 | 12 - 13 | 5084 | 63 | ||
More Information
Set Membership: |
|

Itpr
Genomic Map