Assay Details
| Gene Symbol: Itpr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300251.1 | NP_001287180.1 | 15 - 16 | 8210 | 72 | |
| NM_169060.2 | NP_730941.1 | 16 - 17 | 8192 | 72 | ||
| NM_169061.2 | NP_730942.1 | 15 - 16 | 8165 | 72 | ||
More Information
Set Membership: |
|

Itpr
Genomic Map