Assay Details
Gene Symbol: Pak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001144541.2 | NP_001138013.2 | 3 - 4 | 877 | 88 | |
NM_001275380.2 | NP_001262309.1 | 3 - 4 | 579 | 88 | ||
NM_001275381.2 | NP_001262310.1 | 3 - 4 | 584 | 88 | ||
NM_001275382.1 | NP_001262311.1 | 3 - 4 | 838 | 88 | ||
NM_001275383.1 | NP_001262312.1 | 3 - 4 | 579 | 88 | ||
NM_001275384.1 | NP_001262313.1 | 3 - 4 | 877 | 88 | ||
NM_079942.4 | NP_524681.1 | 3 - 4 | 584 | 88 | ||
NM_169136.3 | NP_731073.1 | 3 - 4 | 877 | 88 | ||
NM_169137.3 | NP_731074.1 | 3 - 4 | 480 | 88 | ||
GenBank mRNA | BT010291.1 | - | 3 - 4 | 845 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
