Assay Details
Gene Symbol: Pak | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275380.2 | NP_001262309.1 | 5 - 6 | 1340 | 80 | |
NM_001275384.1 | NP_001262313.1 | 5 - 6 | 1638 | 80 | ||
NM_079942.4 | NP_524681.1 | 5 - 6 | 1345 | 80 | ||
NM_169136.3 | NP_731073.1 | 5 - 6 | 1638 | 80 | ||
NM_169137.3 | NP_731074.1 | 5 - 6 | 1241 | 80 | ||
GenBank mRNA | BT010291.1 | - | 5 - 6 | 1606 | 80 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
