Assay Details
| Gene Symbol: Cenp-C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300308.1 | NP_001287237.1 | 5 - 6 | 4028 | 117 | |
| NM_169228.3 | NP_731254.2 | 5 - 6 | 4064 | 117 | ||
| GenBank mRNA | BT133292.1 | - | 5 - 6 | 4001 | 117 | |
More Information
Set Membership: |
|

Cenp-C
Genomic Map