Assay Details
| Gene Symbol: Kap-alpha3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170101.1 | NP_001163572.1 | 4 - 5 | 1657 | 94 | |
| NM_169295.3 | NP_731377.1 | 5 - 6 | 1676 | 94 | ||
| NM_169296.3 | NP_731378.1 | 5 - 6 | 1644 | 94 | ||
| NM_176437.2 | NP_788614.1 | 5 - 6 | 1579 | 94 | ||
| GenBank mRNA | AF152928.1 | - | 5 - 6 | 1663 | 94 | |
| AY069430.1 | - | 5 - 6 | 1644 | 94 | ||
More Information
Set Membership: |
|

Kap-alpha3
Genomic Map