Assay Details
Gene Symbol: put | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275645.1 | NP_001262574.1 | 3 - 4 | 863 | 79 | |
NM_001275646.1 | NP_001262575.1 | 3 - 4 | 863 | 79 | ||
NM_169591.3 | NP_731926.1 | 3 - 4 | 863 | 79 | ||
GenBank mRNA | L38495.1 | - | 3 - 4 | 836 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
