Assay Details
| Gene Symbol: Cad88C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169594.3 | NP_731930.3 | 9 - 10 | 5845 | 117 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cad88C| Gene Symbol: Cad88C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169594.3 | NP_731930.3 | 9 - 10 | 5845 | 117 | |
Set Membership: |
|