Assay Details
| Gene Symbol: eff | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001275649.1 | NP_001262578.1 | 4 - 5 | 692 | 73 | |
| NM_001275650.1 | NP_001262579.1 | 4 - 5 | 692 | 73 | ||
| NM_169601.2 | NP_731941.1 | 5 - 6 | 1154 | 73 | ||
| GenBank mRNA | AY060304.1 | - | 5 - 6 | 990 | 73 | |
| KX531702.1 | - | 4 - 5 | 304 | 73 | ||
| KX532146.1 | - | 4 - 5 | 304 | 73 | ||
| X62575.1 | - | 499 | 73 | |||
More Information
Set Membership: |
|

eff
Genomic Map