Assay Details
Gene Symbol: Hel89B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001300422.1 | NP_001287351.1 | 6 - 7 | 5486 | 80 | |
NM_169693.2 | NP_732097.2 | 6 - 7 | 5913 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
