Assay Details
Gene Symbol: Mhcl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001275694.1 | NP_001262623.1 | 7 - 8 | 3846 | 66 | |
NM_169700.3 | NP_732108.2 | 17 - 18 | 6591 | 66 | ||
NM_169703.1 | NP_732111.1 | 6 - 7 | 3909 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
