Assay Details
| Gene Symbol: Akt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001300424.1 | NP_001287353.1 | 6 - 7 | 1817 | 58 | |
| NM_001300425.1 | NP_001287354.1 | 6 - 7 | 2230 | 58 | ||
| NM_169705.2 | NP_732113.3 | 6 - 7 | 2165 | 58 | ||
| NM_169706.2 | NP_732114.1 | 6 - 7 | 2230 | 58 | ||
| NM_169707.2 | NP_732115.1 | 6 - 7 | 2165 | 58 | ||
| GenBank mRNA | AY069856.1 | - | 6 - 7 | 2197 | 58 | |
More Information
Set Membership: |
|

Akt1
Genomic Map