Assay Details
| Gene Symbol: Dhfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001043255.2 | NP_001036720.1 | 1 - 2 | 169 | 63 | |
| NM_169721.2 | NP_732147.1 | 1 - 2 | 102 | 63 | ||
| GenBank mRNA | BT132691.1 | - | 1 - 2 | 118 | 63 | |
More Information
Set Membership: |
|

Dhfr
Genomic Map