Assay Details
| Gene Symbol: Fas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104344.2 | NP_001097814.1 | 12 - 13 | 1875 | 72 | |
| NM_001144593.2 | NP_001138065.1 | 11 - 12 | 1783 | 72 | ||
| NM_001144594.2 | NP_001138066.1 | 11 - 12 | 1836 | 72 | ||
| NM_001275716.1 | NP_001262645.1 | 11 - 12 | 1836 | 72 | ||
| NM_169724.2 | NP_732164.1 | 13 - 14 | 1854 | 72 | ||
| NM_169725.2 | NP_732165.1 | 12 - 13 | 1792 | 72 | ||
| NM_169726.2 | NP_732166.1 | 11 - 12 | 1783 | 72 | ||
| GenBank mRNA | BT044209.1 | - | 11 - 12 | 1814 | 72 | |
| M20545.1 | - | 12 - 13 | 1771 | 72 | ||
More Information
Set Membership: |
|

Fas1
Genomic Map