Assay Details
| Gene Symbol: Fas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104344.2 | NP_001097814.1 | 7 - 8 | 1093 | 72 | |
| NM_001144593.2 | NP_001138065.1 | 6 - 7 | 1001 | 72 | ||
| NM_001144594.2 | NP_001138066.1 | 6 - 7 | 1054 | 72 | ||
| NM_001275716.1 | NP_001262645.1 | 6 - 7 | 1054 | 72 | ||
| NM_169724.2 | NP_732164.1 | 6 - 7 | 1054 | 72 | ||
| NM_169725.2 | NP_732165.1 | 6 - 7 | 1001 | 72 | ||
| NM_169726.2 | NP_732166.1 | 6 - 7 | 1001 | 72 | ||
| GenBank mRNA | BT044209.1 | - | 6 - 7 | 1032 | 72 | |
| M20545.1 | - | 6 - 7 | 980 | 72 | ||
More Information
Set Membership: |
|

Fas1
Genomic Map