Assay Details
| Gene Symbol: Fas1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104344.2 | NP_001097814.1 | 10 - 11 | 1503 | 75 | |
| NM_001144593.2 | NP_001138065.1 | 9 - 10 | 1411 | 75 | ||
| NM_001144594.2 | NP_001138066.1 | 9 - 10 | 1464 | 75 | ||
| NM_001275716.1 | NP_001262645.1 | 9 - 10 | 1464 | 75 | ||
| NM_169724.2 | NP_732164.1 | 11 - 12 | 1482 | 75 | ||
| NM_169725.2 | NP_732165.1 | 10 - 11 | 1420 | 75 | ||
| NM_169726.2 | NP_732166.1 | 9 - 10 | 1411 | 75 | ||
| GenBank mRNA | BT044209.1 | - | 9 - 10 | 1442 | 75 | |
| M20545.1 | - | 10 - 11 | 1399 | 75 | ||
More Information
Set Membership: |
|

Fas1
Genomic Map