Assay Details
Gene Symbol: Dscam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104355.2 | NP_001097825.1 | 30 - 31 | 6138 | 65 | |
NM_169761.2 | NP_732242.2 | 30 - 31 | 5661 | 65 | ||
NM_206504.2 | NP_996226.2 | 30 - 31 | 5661 | 65 |

More Information
Set Membership: |
![]() ![]() |
Gene Ontology Categories:
