Assay Details
Gene Symbol: Dscam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104355.2 | NP_001097825.1 | 8 - 9 | 1575 | 55 | |
NM_001104356.2 | NP_001097826.1 | 7 - 8 | 1170 | 55 | ||
NM_169761.2 | NP_732242.2 | 7 - 8 | 1170 | 55 | ||
NM_206504.2 | NP_996226.2 | 7 - 8 | 1170 | 55 | ||
GenBank mRNA | BT014656.1 | - | 7 - 8 | 1118 | 55 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
