Assay Details
Gene Symbol: Dscam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104355.2 | NP_001097825.1 | 10 - 11 | 1825 | 83 | |
NM_001104356.2 | NP_001097826.1 | 9 - 10 | 1420 | 83 | ||
NM_169761.2 | NP_732242.2 | 9 - 10 | 1420 | 83 | ||
NM_206504.2 | NP_996226.2 | 9 - 10 | 1420 | 83 | ||
GenBank mRNA | BT014656.1 | - | 9 - 10 | 1368 | 83 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
