Assay Details
Gene Symbol: Rpb4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014633.3 | NP_001014633.2 | 2 - 3 | 353 | 97 | |
GenBank mRNA | BT023308.1 | - | 2 - 3 | 316 | 97 | |
BT023309.1 | - | 2 - 3 | 300 | 97 |
Gene Symbol: Ada2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014634.3 | NP_001014634.1 | 4 - 5 | 2016 | 97 | |
NM_001014635.3 | NP_001014635.1 | 3 - 4 | 2031 | 97 | ||
NM_001014636.3 | NP_001014636.1 | 3 - 4 | 2076 | 97 | ||
NM_001014637.3 | NP_001014637.1 | 4 - 5 | 1971 | 97 | ||
NM_001170174.2 | NP_001163645.1 | 5 - 6 | 1901 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
