Assay Details
Gene Symbol: GluClalpha | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038972.4 | NP_001034061.2 | 3 - 4 | 832 | 115 | |
NM_001170184.2 | NP_001163655.1 | 3 - 4 | 832 | 115 | ||
NM_001300479.1 | NP_001287408.1 | 3 - 4 | 832 | 115 | ||
NM_001300480.1 | NP_001287409.1 | 3 - 4 | 832 | 115 | ||
NM_001300481.1 | NP_001287410.1 | 3 - 4 | 832 | 115 | ||
NM_169873.4 | NP_732447.2 | 3 - 4 | 832 | 115 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
