Assay Details
Gene Symbol: CG31176 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_169963.2 | NP_732650.1 | 1 - 2 | 1347 | 113 | |
GenBank mRNA | BT056250.1 | - | 1 - 2 | 1345 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
