Assay Details
| Gene Symbol: CG31176 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_169963.2 | NP_732650.1 | 2 - 3 | 1754 | 111 | |
| GenBank mRNA | BT056250.1 | - | 2 - 3 | 1752 | 111 | |
More Information
Set Membership: |
|

CG31176
Genomic Map