Assay Details
| Gene Symbol: Efa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104403.2 | NP_001097873.1 | 14 - 15 | 4779 | 85 | |
| NM_001104404.2 | NP_001097874.1 | 13 - 14 | 5181 | 85 | ||
| NM_176541.3 | NP_788718.3 | 13 - 14 | 6560 | 85 | ||
More Information
Set Membership: |
|

Efa6
Genomic Map