Assay Details
| Gene Symbol: Efa6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001104403.2 | NP_001097873.1 | 9 - 10 | 3942 | 81 | |
| NM_001104404.2 | NP_001097874.1 | 8 - 9 | 4344 | 81 | ||
| NM_001170220.2 | NP_001163691.2 | 9 - 10 | 4584 | 81 | ||
| NM_170027.4 | NP_732769.3 | 9 - 10 | 3942 | 81 | ||
| NM_176541.3 | NP_788718.3 | 8 - 9 | 4344 | 81 | ||
More Information
Set Membership: |
|

Efa6
Genomic Map