Assay Details
| Gene Symbol: Gdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_079746.6 | NP_524470.4 | 2 - 3 | 947 | 73 | |
| NM_170101.3 | NP_732922.1 | 2 - 3 | 587 | 73 | ||
| NM_206550.2 | NP_996273.2 | 2 - 3 | 587 | 73 | ||
| NM_206551.3 | NP_996274.1 | 2 - 3 | 947 | 73 | ||
| GenBank mRNA | AY061323.1 | - | 2 - 3 | 925 | 73 | |
| BT128894.1 | - | 2 - 3 | 582 | 73 | ||
More Information
Set Membership: |
|

Gdh
Genomic Map