Assay Details
Gene Symbol: Gdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_079746.6 | NP_524470.4 | 4 - 5 | 1472 | 82 | |
NM_170101.3 | NP_732922.1 | 4 - 5 | 1112 | 82 | ||
GenBank mRNA | AY061323.1 | - | 4 - 5 | 1450 | 82 | |
BT128894.1 | - | 4 - 5 | 1107 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
