Assay Details
Gene Symbol: jar | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014647.3 | NP_001014647.2 | 4 - 5 | 734 | 70 | |
NM_001014648.3 | NP_001014648.2 | 4 - 5 | 734 | 70 | ||
NM_001014649.3 | NP_001014649.2 | 4 - 5 | 734 | 70 | ||
NM_001014650.3 | NP_001014650.2 | 4 - 5 | 734 | 70 | ||
NM_001275976.1 | NP_001262905.1 | 4 - 5 | 492 | 70 | ||
NM_001275977.2 | NP_001262906.1 | 4 - 5 | 1201 | 70 | ||
NM_001300572.1 | NP_001287501.1 | 4 - 5 | 734 | 70 | ||
NM_079754.4 | NP_524478.4 | 4 - 5 | 734 | 70 | ||
NM_170138.3 | NP_732976.1 | 4 - 5 | 492 | 70 | ||
GenBank mRNA | BT133083.1 | - | 4 - 5 | 731 | 70 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
