Assay Details
Gene Symbol: slo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014654.3 | NP_001014654.1 | 14 - 15 | 2664 | 89 | |
NM_170164.4 | NP_733029.1 | 15 - 16 | 2405 | 89 | ||
NM_206566.4 | NP_996289.1 | 15 - 16 | 2405 | 89 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
