Assay Details
Gene Symbol: slo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014651.3 | NP_001014651.1 | 16 - 17 | 2733 | 76 | |
NM_001014654.3 | NP_001014654.1 | 17 - 18 | 2808 | 76 | ||
NM_001014655.3 | NP_001014655.1 | 16 - 17 | 2733 | 76 | ||
NM_001014656.3 | NP_001014656.1 | 16 - 17 | 2769 | 76 | ||
NM_001014659.3 | NP_001014659.1 | 18 - 19 | 2847 | 76 | ||
NM_001014660.3 | NP_001014660.1 | 16 - 17 | 2733 | 76 | ||
NM_001014661.4 | NP_001014661.2 | 18 - 19 | 2564 | 76 | ||
NM_001014662.3 | NP_001014662.1 | 16 - 17 | 2733 | 76 | ||
NM_001014663.3 | NP_001014663.1 | 16 - 17 | 2733 | 76 | ||
NM_001014664.3 | NP_001014664.1 | 16 - 17 | 2733 | 76 | ||
NM_001170241.3 | NP_001163712.1 | 19 - 20 | 3092 | 76 | ||
NM_001275995.1 | NP_001262924.1 | 19 - 20 | 2774 | 76 | ||
NM_001275996.1 | NP_001262925.1 | 16 - 17 | 2733 | 76 | ||
NM_079762.4 | NP_524486.2 | 16 - 17 | 2733 | 76 | ||
NM_170164.4 | NP_733029.1 | 18 - 19 | 2549 | 76 | ||
NM_206566.4 | NP_996289.1 | 18 - 19 | 2549 | 76 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
