Assay Details
| Gene Symbol: HDAC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001260367.1 | NP_001247296.1 | 1 - 2 | 237 | 78 | |
| NM_170173.3 | NP_733048.1 | 1 - 2 | 237 | 78 | ||
| GenBank mRNA | BT082053.1 | - | 1 - 2 | 237 | 78 | |
More Information
Set Membership: |
|

HDAC11
Genomic Map