Assay Details
| Gene Symbol: PIG-S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170181.3 | NP_733060.1 | 5 - 6 | 779 | 68 | |
| GenBank mRNA | AY061834.1 | - | 5 - 6 | 751 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIG-S| Gene Symbol: PIG-S | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170181.3 | NP_733060.1 | 5 - 6 | 779 | 68 | |
| GenBank mRNA | AY061834.1 | - | 5 - 6 | 751 | 68 | |
Set Membership: |
|