Assay Details
Gene Symbol: gro | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001202364.1 | NP_001189293.1 | 6 - 7 | 1094 | 68 | |
NM_001202365.2 | NP_001189294.1 | 7 - 8 | 1115 | 68 | ||
NM_001260380.2 | NP_001247309.1 | 7 - 8 | 1115 | 68 | ||
NM_001276052.1 | NP_001262981.1 | 6 - 7 | 1082 | 68 | ||
NM_001300610.1 | NP_001287539.1 | 6 - 7 | 1082 | 68 | ||
NM_079790.5 | NP_524514.2 | 6 - 7 | 1027 | 68 | ||
NM_170254.3 | NP_733133.1 | 6 - 7 | 1107 | 68 | ||
NM_170255.4 | NP_733134.1 | 6 - 7 | 1088 | 68 | ||
NM_170256.2 | NP_733135.1 | 6 - 7 | 1006 | 68 | ||
NM_206575.3 | NP_996298.1 | 7 - 8 | 1121 | 68 | ||
GenBank mRNA | AF145695.1 | - | 6 - 7 | 1073 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
